missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lithium tri-tert-butoxyaluminum hydride, 94%

Manufacturer:  Alfa Aesar™ 3071609

 View more versions of this product

Catalog No. AA3071609



Specifications

Specifications

Lithium tri-tert-butoxyaluminum hydride
17476-04-9
1.03g/mL
Odorless
Li[(CH3)3CO]3AlH
UN1409
Lithium hydridotris(tert-butoxy)aluminate
Powder
~319°C (decomposition)
10g
Gray to White
Moisture Sensitive
MFCD00011532
5796791
254.28
94%

Chemical Identifiers

17476-04-9
MFCD00011532
Lithium hydridotris(tert-butoxy)aluminate
Safety and Handling

Safety and Handling

GHS H Statement
H228-H261-H314-H318
Flammable solid.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P210-P280-P305+P351+P338-P310-P370+P378i-P402+P404
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
In case of fire: Use for extinction: Fire-extinguishing powder.
Store in a dry place.
Store in a closed container.

Danger

DOTInformation : Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: METAL HYDRIDES, WATER REACTIVE, N.O.S.

RTECSNumber : OJ5585000

TSCA : TSCA

SDS Documents