missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lithium tri-tert-butoxyaluminum hydride, 94%

$110.70 - $110.70

Chemical Identifiers

CAS 17476-04-9
Linear Formula Li[(CH3)3CO]3AlH
Formula Weight 254.28
MDL Number MFCD00011532
UN Number UN1409
Beilstein 5796791
Synonym Lithium hydridotris(tert-butoxy)aluminate
Chemical Name or Material Lithium tri-tert-butoxyaluminum hydride
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3071609 SDS Alfa Aesar™
3071609
10g Each for $110.70
Specifications Description & Specifications

Chemical Identifiers

17476-04-9
254.28
UN1409
Lithium hydridotris(tert-butoxy)aluminate
Li[(CH3)3CO]3AlH
MFCD00011532
5796791
Lithium tri-tert-butoxyaluminum hydride

Specifications

≈319°C (dec.)
10g
White to pale gray
Moisture Sensitive
Li[(CH3)3CO]3AlH
MFCD00011532
5796791
Lithium tri-tert-butoxyaluminum hydride
17476-04-9
1.03g/mL
Odorless
Powder
254.28
UN1409
Lithium hydridotris(tert-butoxy)aluminate
94%

Safety and Handling

GHS H Statement
H228-H261-H314-H318
Flammable solid.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P210-P280-P305+P351+P338-P310-P370+P378i-P402+P404
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
In case of fire: Use for extinction: Fire-extinguishing powder.
Store in a dry place.
Store in a closed container.

Danger

DOTInformation : Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: METAL HYDRIDES, WATER REACTIVE, N.O.S.

RTECSNumber : OJ5585000

TSCA : TSCA