Learn More
Lithium tri-tert-butoxyaluminohydride, 93-98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 125160250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentSpecifications
Lithium tri-tert-butoxyaluminohydride | |
17476-04-9 | |
Gray-White to White | |
Authentic | |
Glass Bottle packed in tin can | |
LiAI[OC(CH3)3]3H | |
25 g | |
Solubility in water: reacts violently. Other solubilities: 36g/100g thf (25°C),2g/100g diethyl ether (25°C),41g/100g diglyme (25°C) | |
254.27 | |
93 to 98% |
93-98% | |
319.0°C | |
0.9 | |
93% min | |
C12H28AlLiO3 | |
MFCD00011532 | |
01,620; 02,251; 03,188; 04,312; 10,248; 12,286; 13,342; 16,197 | |
280 °C | |
254.27 | |
Powder |
Chemical Identifiers
17476-04-9 | |
C12H28AlLiO3 | |
254.27 | |
MFCD00011532 |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.
Reacts violently with water.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 241-490-8
RUO – Research Use Only