missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium tri-tert-butoxyaluminohydride, 93-98%, Thermo Scientific™

$154.86 - $154.86

Chemical Identifiers

CAS 17476-04-9
Molecular Formula C12H28AlLiO3
Molecular Weight (g/mol) 254.27
MDL Number MFCD00011532
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC125160250
SDS View Documents Promotion Details
Thermo Fisher Scientific
125160250
25g
Each for $154.86
 
AC125161000
SDS View Documents Promotion Details
Thermo Fisher Scientific
125161000
100g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Reduction reagent
Specifications

Chemical Identifiers

17476-04-9
C12H28AlLiO3
254.27
MFCD00011532

Specifications

17476-04-9
Gray-White to White
93% min
Authentic
MFCD00011532
01,620; 02,251; 03,188; 04,312; 10,248; 12,286; 13,342; 16,197
Solubility in water: reacts violently. Other solubilities: 36g/100g thf (25°C),2g/100g diethyl ether (25°C),41g/100g diglyme (25°C)
280 °C
Powder
Lithium tri-tert-butoxyaluminohydride, 93-98%
0.9
319.0°C
C12H28AlLiO3
LiAI[OC(CH3)3]3H
Glass Bottle packed in tin can
25g
254.27
254.27
93 to 98%
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.
Reacts violently with water.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 241-490-8

SDS
Product Certifications

RUO – Research Use Only