Please login to your online account to display your discounted pricing

Lithium tri-sec-butylborohydride, 1M solution in THF, AcroSeal™, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Lithium tri-sec-butylborohydride
CAS 38721-52-7
CAS 109-99-9
CAS 1113-78-6
Molecular Formula C12H28BLi
Linear Formula LiB[CH(CH3)C2H5]3H
Formula Weight 190.11
MDL Number MFCD00011708
Synonym Lithium triisobutylhydroborate
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC176458000

  SDS
thermo fisher scientific
176458000
AcroSeal™ Glass Bottle 800mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC176451000

  SDS
thermo fisher scientific
176451000
AcroSeal™ Glass Bottle in Tin Can 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Formula Weight 190.11
Quantity 800mL
Color Clear colorless
Packaging AcroSeal™ Glass Bottle
EINECS Number 254-101-1
TSCA TSCA
Density 0.8900g/mL
Fieser 04,312; 06,348; 07,307; 08,308; 10,248; 12,286; 14,195; 15,192
Merck Index 15, 8564
Chemical Name or Material Lithium tri-sec-butylborohydride
Molecular Formula C12H28BLi
Linear Formula LiB[CH(CH3)C2H5]3H
MDL Number MFCD00011708
CAS 38721-52-7
Synonym Lithium triisobutylhydroborate
Health Hazard 1 Danger
Health Hazard 2 GHS P Statement Causes severe skin burns and eye damage. In contact with water releases flammable gases which may ignite spontaneously. Highly flammable liquid and vapor. May cause respiratory irritation. Reacts viole
CAS 109-99-9
Health Hazard 3 GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Store in a dry place. Store in a closed container. Avoid breathing dust/fume/gas/mist/vapors/spray. IF IN EYES: Rinse cautiously with
CAS 1113-78-6

Reduction reagent

Safety and Handling

Danger

GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Store in a dry place. Store in a closed container. Avoid breathing dust/fume/gas/mist/vapors/spray. IF IN EYES: Rinse cautiously with

GHS P Statement Causes severe skin burns and eye damage. In contact with water releases flammable gases which may ignite spontaneously. Highly flammable liquid and vapor. May cause respiratory irritation. Reacts viole