Please login to your online account to display your discounted pricing

Alfa Aesar™ Lithium tri-sec-butylborohydride, 1.0M in THF

As a selective reducing agent

Chemical Identifiers

Chemical Name or Material Lithium tri-sec-butylborohydride
CAS 38721-52-7
Molecular Formula C12H28BLi
Linear Formula LiB[CH(CH3)CH2CH3]3H
Formula Weight 190.11
MDL Number MFCD00011708
UN Number UN3394
Beilstein 4007327
Merck Index 14,8427
Synonym Lithium tri-sec-butylhydridoborate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA43845AC

 
alfa aesar™
43845AC
25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA43845AE

 
alfa aesar™
43845AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 190.11
Chemical Name or Material Lithium tri-sec-butylborohydride
TSCA TSCA
Physical Form Liquid
Molecular Formula C12H28BLi
Linear Formula LiB[CH(CH3)CH2CH3]3H
Density 0.870g/mL
MDL Number MFCD00011708
CAS 38721-52-7
UN Number UN3394
Beilstein 4007327
Merck Index 14,8427
Synonym Lithium tri-sec-butylhydridoborate
Concentration 1.0M in THF
Color Colorless to yellow
Health Hazard 1 Danger
Quantity 25mL
Flash Point −17°C (1°F)
Sensitivity Moisture Sensitive
Health Hazard 2 GHS H Statement
H225-H250-H260-H314-H318
Highly flammable liquid and vapor.
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P210-P280-P305+P351+P338-P310-P370+P378i-P402+P404
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
In case of fire: Use for extinction: Fire-extinguishing powder.
Store in a dry place.
Store in a closed container.
DOT Information Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, PYROPHORIC, WATER-REACTIVE
Safety and Handling

Danger

GHS P Statement
P210-P280-P305+P351+P338-P310-P370+P378i-P402+P404
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
In case of fire: Use for extinction: Fire-extinguishing powder.
Store in a dry place.
Store in a closed container.

GHS H Statement
H225-H250-H260-H314-H318
Highly flammable liquid and vapor.
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
Causes serious eye damage.