Please login to your online account to display your discounted pricing

Lithium thiocyanate hydrate, 98+%, extra pure, ACROS Organics™


Quantity 25g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC388660250

 View more versions of this product

Catalog No. AC388660250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade Extra Pure
Physical Form Crystalline Mass or Crystals
Formula Weight 65.02
Quantity 25g
Color Colorless
Assay 98.0 to 102.0% (on dry substance)
Chemical Name or Material Lithium thiocyanate hydrate
Insoluble in Water 0.005% max. (5% max. solution)
Iron (Fe) 2ppm max.
Assay Percent Range 98+%
MDL Number MFCD00149773
CAS 123333-85-7
Water 21 to 36% (K.F.)
Health Hazard 1 Warning
Packaging Plastic bottle
Reducing Substances 10mg iodine per 100 gram max.
Molecular Formula CLiNS·xH2O
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful to aquatic life with long lasting effects.
Harmful in contact with skin.
Contact with acids liberates very toxic gas.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
Keep container tightly closed.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Lithium thiocyanate hydrate
CAS 123333-85-7
Purity Grade Extra Pure
Assay 98.0 to 102.0% (on dry substance)
Assay Percent Range 98+%
Molecular Formula CLiNS·xH2O
Formula Weight 65.02
MDL Number MFCD00149773
Accessories

Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
Keep container tightly closed.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful to aquatic life with long lasting effects.
Harmful in contact with skin.
Contact with acids liberates very toxic gas.

SDS & Safety Documents