Please login to your online account to display your discounted pricing

Lithium thiocyanate hydrate, 98+%, extra pure, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Lithium thiocyanate hydrate
CAS 123333-85-7
Purity Grade Extra pure
Assay 98.0 to 102.0% (on dry substance)
Assay Percent Range 98+%
Molecular Formula CLiNS·xH2O
Formula Weight 65.02g/mol
MDL Number MFCD00149773
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC388660250

  SDS
thermo fisher scientific
388660250
Plastic bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Insoluble in Water 0.005% max. (5% max. solution)
Iron (Fe) 2ppm max.
Packaging Plastic bottle
Reducing Substances 10mg iodine per 100 gram max.
Water 21 to 36% (K.F.)
Color Colorless
Quantity 25g
Chemical Name or Material Lithium thiocyanate hydrate
CAS 123333-85-7
Purity Grade Extra pure
Physical Form Colorless crystalline mass or crystals
Assay 98.0 to 102.0% (on dry substance)
Assay Percent Range 98+%
Molecular Formula CLiNS·xH2O
Formula Weight 65.02g/mol
MDL Number MFCD00149773
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful to aquatic life with long lasting effects.
Harmful in contact with skin.
Contact with acids liberates very toxic gas.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
Keep container tightly closed.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
Keep container tightly closed.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful to aquatic life with long lasting effects.
Harmful in contact with skin.
Contact with acids liberates very toxic gas.