Please login to your online account to display your discounted pricing

Alfa Aesar™ Lithium thiocyanate hydrate

$151.16 - $151.16

Chemical Identifiers

Chemical Name or Material Lithium thiocyanate hydrate
CAS 123333-85-7
Formula Weight 65.02 (Anhydrous)
MDL Number MFCD00149773
Synonym Lithium rhodanide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1134918

 
alfa aesar™
1134918*
50g Each for $151.16
Description & Specifications

Specifications

Formula Weight 65.02 (Anhydrous)
Health Hazard 3 GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Lithium thiocyanate hydrate
TSCA TSCA
Physical Form Crystalline
Sensitivity Hygroscopic
MDL Number MFCD00149773
CAS 123333-85-7
Synonym Lithium rhodanide
Quantity 50g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
RTECS Number XL1540000
Safety and Handling

Warning

GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.