missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium Tetrakis(pentafluorophenyl)borate - Ethyl Ether Complex, TCI America™

Manufacturer:  TCI America L01585G

 View more versions of this product

Catalog No. L01585G



Specifications

Specifications

Lithium Tetrakis(pentafluorophenyl)borate - Ethyl Ether Complex
5g
C28H10BF20LiO
3175
[Li+].[B-](C1=C(C(=C(C(=C1F)F)F)F)F)(C2=C(C(=C(C(=C2F)F)F)F)F)(C3=C(C(=C(C(=C3F)F)F)F)F)C4=C(C(=C(C(=C4F)F)F)F)F.CCOCC
760.105
Crystal-Powder at 20°C
Undesignated
155543-02-5
MFCD01861289
KPLZKJQZPFREPG-UHFFFAOYSA-N
lithium;ethoxyethane;tetrakis(2,3,4,5,6-pentafluorophenyl)boranuide
23701484
≥70.0% (T)

Chemical Identifiers

155543-02-5
760.105
KPLZKJQZPFREPG-UHFFFAOYSA-N
lithium;ethoxyethane;tetrakis(2,3,4,5,6-pentafluorophenyl)boranuide
C28H10BF20LiO
MFCD01861289
23701484
[Li+].[B-](C1=C(C(=C(C(=C1F)F)F)F)F)(C2=C(C(=C(C(=C2F)F)F)F)F)(C3=C(C(=C(C(=C3F)F)F)F)F)C4=C(C(=C(C(=C4F)F)F)F)F.CCOCC
Safety and Handling

Safety and Handling

TSCA : No

Documents