Please login to your online account to display your discounted pricing

Alfa Aesar™ Lithium tetrafluoroborate, ultra dry, 99.997% (metals basis)

Chemical Identifiers

Chemical Name or Material Lithium tetrafluoroborate
CAS 14283-07-9
Name Note Ultra dry
Linear Formula LiBF4
Formula Weight 93.75
MDL Number MFCD00011087
UN Number UN3260
Merck Index 14,5543
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1453206

  SDS
alfa aesar™
1453206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1453214

  SDS
alfa aesar™
1453214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Lithium tetrafluoroborate
Name Note Ultra dry
CAS 14283-07-9
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H302-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1
  • Ampouled under argon
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
TSCA TSCA
Physical Form Powder
Assay 99.997%
Assay Percent Notes (metals basis)
Linear Formula LiBF4
Formula Weight 93.75
MDL Number MFCD00011087
UN Number UN3260
Merck Index 14,5543
Odor Odorless
Packaging Ampule
Sensitivity Hygroscopic
Color White to off-white
Melting Point 293° to 300°C (dec.)
Quantity 5g

  • Ampouled under argon

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H302-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.

  • Ampouled under argon