missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lithium tetrafluoroborate, ultra dry, 99.997% (metals basis)

Manufacturer:  Alfa Aesar™ 1453214

 View more versions of this product

Catalog No. AA1453214



Description

Description

  • Ampouled under argon
Specifications

Specifications

Lithium tetrafluoroborate
14283-07-9
BF4Li
MFCD00011087
14,5543
UFXJWFBILHTTET-UHFFFAOYSA-N
lithium;tetrafluoroborate
4298216
Powder
Odorless
Hygroscopic
293°C to 300°C (decomposition)
Ultra dry
(metals basis)
LiBF4
UN3260
lithium tetrafluoroborate, lithium fluoroborate, lithium tetra fluoroborate, lithium 1+ ion tetrafluoroborate, lithium borofluoride, unii-af751cnk2n, libf4, lithotab tetrafluoroborate, acmc-209cp3, af751cnk2n
[Li+].[B-](F)(F)(F)F
93.744
93.75
99.997%
Ampule
White
25g

Chemical Identifiers

14283-07-9
93.744
UFXJWFBILHTTET-UHFFFAOYSA-N
4298216
[Li+].[B-](F)(F)(F)F
BF4Li
MFCD00011087
lithium tetrafluoroborate, lithium fluoroborate, lithium tetra fluoroborate, lithium 1+ ion tetrafluoroborate, lithium borofluoride, unii-af751cnk2n, libf4, lithotab tetrafluoroborate, acmc-209cp3, af751cnk2n
lithium;tetrafluoroborate
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

TSCA : TSCA

storageNote1 :

  • Ampouled under argon

Documents