Learn More
Lithium tetrafluoroborate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01152809
Description
Lithium tetrafluoroborate is extensively utilized as an electrolyte in primary and secondary cells. It is used in rechargeable Li ion batteries. The electrolytes are non aqueous solutions of lithium tetrafluoroborate in the organic media.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Lithium tetrafluoroborate | |
14283-07-9 | |
Powder | |
BF4Li | |
MFCD00011087 | |
14,5543 | |
Soluble in water and tetrahydrofuran. Slightly soluble is diethyl ether. | |
[Li+].F[B-](F)(F)F | |
93.74 | |
93.75 | |
Odorless |
293°C to 300°C (decomposition) | |
White | |
10 g | |
LiBF4 | |
UN3260 | |
lithium tetrafluoroborate, lithium fluoroborate, lithium tetra fluoroborate, lithium 1+ ion tetrafluoroborate, lithium borofluoride, unii-af751cnk2n, libf4, lithotab tetrafluoroborate, acmc-209cp3, af751cnk2n | |
UFXJWFBILHTTET-UHFFFAOYSA-N | |
lithium;tetrafluoroborate | |
4298216 | |
98% | |
Very Hygroscopic |
Chemical Identifiers
14283-07-9 | |
93.74 | |
UFXJWFBILHTTET-UHFFFAOYSA-N | |
4298216 | |
[Li+].F[B-](F)(F)F |
BF4Li | |
MFCD00011087 | |
lithium tetrafluoroborate, lithium fluoroborate, lithium tetra fluoroborate, lithium 1+ ion tetrafluoroborate, lithium borofluoride, unii-af751cnk2n, libf4, lithotab tetrafluoroborate, acmc-209cp3, af751cnk2n | |
lithium;tetrafluoroborate |
Safety and Handling
GHS H Statement
H314-H318-H302-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.
P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P362-P501c
H302+H312+H332-H314-H318
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 238-178-9
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only