Learn More
Lithium tetrachloropalladate(II), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01731603
Description
It act as a catalyst in organic reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Lithium tetrachloropalladate(II) | |
Crystalline | |
MFCD00011082 | |
262.09 |
15525-45-8 | |
1 g | |
Soluble in water. | |
Moisture sensitive |
Chemical Identifiers
15525-45-8 | |
MFCD00011082 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 239-567-6
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only