missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium tetrachlorocuprate, 0.1M in THF, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 020620AP

 View more versions of this product

Catalog No. AA20620AP



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Lithium tetrachlorocuprate
15489-27-7
500 mL
MFCD00011081
Difficult to mix.
[Li+].[Li+].Cl[Cu-2](Cl)(Cl)Cl
219.226
219.24
Moisture sensitive
Orange
Liquid
Cl4CuLi2
UN2056
HCJWWBBBSCXJMS-UHFFFAOYSA-J
dilithium;tetrachlorocopper(2-)
11074879
0.1M in THF
0.910 g/mL

Chemical Identifiers

15489-27-7
219.226
HCJWWBBBSCXJMS-UHFFFAOYSA-J
dilithium;tetrachlorocopper(2-)
Cl4CuLi2
MFCD00011081
11074879
[Li+].[Li+].Cl[Cu-2](Cl)(Cl)Cl
Safety and Handling

Safety and Handling

GHS H Statement
H225-H318-H351-H335-H315
Highly flammable liquid and vapor.
Causes serious eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
Causes skin irritation.

P201-P202-P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280i-P281-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P330-P370+P378q-P501c

H225-H302-H319-H335-H336-H351

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: TETRAHYDROFURAN

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only