Please login to your online account to display your discounted pricing

Lithium tetraborate, Puratronic™, 99.998% (metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Lithium tetraborate
CAS 12007-60-2
Name Note Puratronic™
Molecular Formula B4Li2O7
Linear Formula Li2B4O7
Formula Weight 169.12
MDL Number MFCD00011083
Merck Index 14,5524
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA1074422

 
alfa aesar
1074422
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 169.12
Quantity 100g
Chemical Name or Material Lithium tetraborate
TSCA TSCA
Mesh Size -22 Mesh
Odor Odorless
Name Note Puratronic™
Physical Form Powder
Assay 99.998%
Assay Percent Notes (metals basis)
Molecular Formula B4Li2O7
Linear Formula Li2B4O7
MDL Number MFCD00011083
CAS 12007-60-2
Merck Index 14,5524
Color White
Melting Point 930°C
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335;
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a;
Avoid breathing dust/fume/gas/mist/vapours/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Density 2.4
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a;
Avoid breathing dust/fume/gas/mist/vapours/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335;
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.