Please login to your online account to display your discounted pricing

Lithium Tetraborate, Flux Grade, BAKER INSTRA-ANALYZED™ Reagent, J.T.Baker™

Acids and standards that are purified and characterized for use in trace element analysis in the low parts-per-billion (ppb) range

$726.76 - $726.76

Chemical Identifiers

Chemical Name or Material Lithium Tetraborate
CAS 12007-60-2
Name Note Flux Grade
Purity Grade BAKER INSTRA-ANALYZED™ Reagent
Molecular Formula Li2B4O7
Formula Weight 169.11
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003223

 
avantor performance
450301
Poly Bottle 500g Each for $726.76
Description & Specifications

Specifications

Formula Weight 169.11
Chemical Name or Material Lithium Tetraborate
CAS 12007-60-2
Name Note Flux Grade
Packaging Poly Bottle
Purity Grade BAKER INSTRA-ANALYZED™ Reagent
Specific Gravity 1L = 2.40kg
Molecular Formula Li2B4O7
Quantity 500g