Please login to your online account to display your discounted pricing

Alfa Aesar™ Lithium tetra(2-methyl-8-hydroxyquinolinato)boron, 98%

Chemical Identifiers

Chemical Name or Material Lithium tetra(2-methyl-8-hydroxyquinolinato)boron
CAS 338949-42-1
Formula Weight 650.46
MDL Number MFCD03093696
Synonym Lithium tetrakis[(2-methyl-8-quinolinyl)oxy]borate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH55454MC

 
alfa aesar™
H55454MC
100mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH55454ME

 
alfa aesar™
H55454ME
500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 650.46
Melting Point 250° to 258°C
Chemical Name or Material Lithium tetra(2-methyl-8-hydroxyquinolinato)boron
Physical Form Solid
Assay 98%
MDL Number MFCD03093696
CAS 338949-42-1
Synonym Lithium tetrakis[(2-methyl-8-quinolinyl)oxy]borate
Color White to off-white
Quantity 100mg