Please login to your online account to display your discounted pricing

Lithium Tet/LiMet/LiBromide 49.75%/49.75%/0.50%, SPEX CertiPrep™

$801.49 - $801.49

Chemical Identifiers

Name Note Fusion Flux
Purity Grade Pure
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

FFB500502

 
spex certiprep llc
SPXFFB500502
Triple-leached LDPE bottle 1kg Each for $801.49
Description & Specifications

Specifications

Quantity 1kg
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Chemical Name or Material Lithium Tetraborate/Lithium Metaborate/Lithium Bromide
Recommended Storage Ambient
Packaging Triple-leached LDPE bottle
DOT Information Nonhazardous
Purity Grade Pure
Name Note Fusion Flux

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

Ambient