missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Lithium tert-butoxide, 0.9M (8 wt.%) solution in 2-MeTHF, AcroSeal™, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC446848000
Specifications
Lithium tert-butoxide | |
C4H9LiO | |
2-methyl-2-propanol lithium salt, 2-propanol, 2-methyl-, lithium salt, lithium 1+ ion 2-methylpropan-2-olate, lithium 2-methylpropan-2-olate, lithium t-butoxide, lithium tert-butanolate, lithium tert-butoxide, lithium tert-butylate, tert-butoxylithium, tert-butyl alcohol, lithium salt | |
[Li+].CC(C)(C)[O-] | |
80.05 | |
80.05 | |
0.86 | |
-11°C |
1907-33-1,96-47-9 | |
LiOCH(CH3)3 | |
LZWQNOHZMQIFBX-UHFFFAOYSA-N | |
lithium;2-methylpropan-2-olate | |
23664764 | |
0.8600g/mL | |
800mL |
Chemical Identifiers
1907-33-1 | |
80.05 | |
2-methyl-2-propanol lithium salt, 2-propanol, 2-methyl-, lithium salt, lithium 1+ ion 2-methylpropan-2-olate, lithium 2-methylpropan-2-olate, lithium t-butoxide, lithium tert-butanolate, lithium tert-butoxide, lithium tert-butylate, tert-butoxylithium, tert-butyl alcohol, lithium salt | |
lithium;2-methylpropan-2-olate |
C4H9LiO | |
LZWQNOHZMQIFBX-UHFFFAOYSA-N | |
23664764 | |
[Li+].CC(C)(C)[O-] |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Reacts violently with water.
May form explosive peroxides.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove conta
GHS Signal Word: Danger
EINECSNumber : 217-611-5