missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lithium tert-butoxide, 99%

Manufacturer:  Alfa Aesar™ 46366A1

 View more versions of this product

Catalog No. AA46366A1



Specifications

Specifications

Lithium tert-butoxide
1907-33-1
White to Cream
Air & Moisture Sensitive
C4H9LiO
MFCD00050479
3620018
[Li+].CC(C)(C)[O-]
80.055
80.06
~150°C (decomposition)
1kg
Odorless
99%
(CH3)3COLi
UN3206
LZWQNOHZMQIFBX-UHFFFAOYSA-N
lithium;2-methylpropan-2-olate
23664764
Powder

Chemical Identifiers

1907-33-1
80.055
LZWQNOHZMQIFBX-UHFFFAOYSA-N
lithium;2-methylpropan-2-olate
C4H9LiO
MFCD00050479
23664764
[Li+].CC(C)(C)[O-]
Safety and Handling

Safety and Handling

GHS H Statement
H251-H314-H302
Self-heating: may catch fire.
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement
P210-P280-P305+P351+P338-P310-P370+P378i-P402+P404
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
In case of fire: Use for extinction: Fire-extinguishing powder.
Store in a dry place.
Store in a closed container.

Danger

DOTInformation : Transport Hazard Class: 4.2; Packing Group: II; Proper Shipping Name: ALKALI METAL ALCOHOLATES, SELF-HEATING, CORROSIVE, N.O.S.

RTECSNumber : UB8520000

TSCA : TSCA

SDS Documents