Learn More
Lithium tert-butoxide, 2.2M solution in THF, AcroSeal™, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 379311000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Lithium tert-butoxide | |
0.8900g/mL | |
AcroSeal™ Glass bottle | |
LiOCH(CH3)3 | |
100mL | |
lithium tert-butoxide, lithium 2-methylpropan-2-olate, tert-butoxylithium, lithium tert-butylate, lithium tert-butanolate, tert-butyl alcohol, lithium salt, lithium t-butoxide, 2-methyl-2-propanol lithium salt, 2-propanol, 2-methyl-, lithium salt, lithium 1+ ion 2-methylpropan-2-olate | |
LZWQNOHZMQIFBX-UHFFFAOYSA-N | |
lithium;2-methylpropan-2-olate | |
23664764 |
1907-33-1,109-99-9 | |
−22°C | |
C4H9LiO | |
MFCD00050479 | |
0.89 | |
Solubility in water: vigorous reaction | |
[Li+].CC(C)(C)[O-] | |
80.06 | |
80.06 |
Chemical Identifiers
1907-33-1 | |
80.06 | |
LZWQNOHZMQIFBX-UHFFFAOYSA-N | |
23664764 | |
[Li+].CC(C)(C)[O-] |
C4H9LiO | |
MFCD00050479 | |
lithium tert-butoxide, lithium 2-methylpropan-2-olate, tert-butoxylithium, lithium tert-butylate, lithium tert-butanolate, tert-butyl alcohol, lithium salt, lithium t-butoxide, 2-methyl-2-propanol lithium salt, 2-propanol, 2-methyl-, lithium salt, lithium 1+ ion 2-methylpropan-2-olate | |
lithium;2-methylpropan-2-olate |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
May form explosive peroxides.
Reacts violently with water.
May cause respiratory irritation.
Suspected of causing cancer.<br/
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p
GHS Signal Word: Danger
EINECSNumber : 217-611-5
RUO â Research Use Only