Please login to your online account to display your discounted pricing

Lithium tert-butoxide, 1M solution in hexanes, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Lithium tert-butoxide
Name Note 1M solution in hexanes
CAS 1907-33-1
CAS 110-54-3
Molecular Formula C4H9LiO
Linear Formula LiOCH(CH3)3
Formula Weight 80.06
MDL Number MFCD00050479
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC380158000

  SDS
thermo fisher scientific
AC380158000
AcroSeal™ Glass Bottle 800mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC380151000

  SDS
thermo fisher scientific
AC380151000
AcroSeal™ Glass Bottle 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form May Contain Some Turbidity or Precipitate
Formula Weight 80.06
Quantity 800mL
Packaging AcroSeal™ Glass Bottle
Concentration 0.95 to 1.10M
Density 0.69g/mL
Linear Formula LiOCH(CH3)3
Chemical Name or Material Lithium tert-butoxide
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
Store in a dry place. Store in a closed container.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 1M solution in hexanes
Appearance of Solution Clear yellow solution
CAS 1907-33-1
Health Hazard 1 GHS Signal Word: Danger
CAS 110-54-3
Health Hazard 2 GHS H Statement:
May cause drowsiness or dizziness.
Toxic to aquatic life with long lasting effects.
Causes severe skin burns and eye damage.
May cause damage to organs through prolonged or repeated exposure.
May be fatal if swallowed and enters airways.
Suspected of dama
Molecular Formula C4H9LiO
MDL Number MFCD00050479
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
Store in a dry place. Store in a closed container.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause drowsiness or dizziness.
Toxic to aquatic life with long lasting effects.
Causes severe skin burns and eye damage.
May cause damage to organs through prolonged or repeated exposure.
May be fatal if swallowed and enters airways.
Suspected of dama