missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lithium tartrate monohydrate, 99%

$234.86 - $234.86

Chemical Identifiers

CAS 6108-32-3
Formula Weight 179.97 (161.97 Anhydrous)
MDL Number MFCD00054421
Synonym Tartaric acid lithium salt monohydrate
Chemical Name or Material Lithium tartrate monohydrate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3935718 SDS Alfa Aesar™
3935718
50g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA3935730 SDS Alfa Aesar™
3935730
250g Each for $234.86
Specifications Description & Specifications

Chemical Identifiers

6108-32-3
MFCD00054421
Lithium tartrate monohydrate
179.97 (161.97 Anhydrous)
Tartaric acid lithium salt monohydrate

Specifications

6108-32-3
Odorless
179.97 (161.97 Anhydrous)
Tartaric acid lithium salt monohydrate
Lithium tartrate monohydrate
50g
Powder/Aggregate
MFCD00054421
Soluble in water
99%