missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Lithium tartrate monohydrate, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 03935730
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Lithium tartrate monohydrate | |
Odorless | |
MFCD00054421 | |
lithium tartrate monohydrate, lithium tartrate 1-hydrate, c4h4o6.2li.h2o, dilithium 1+ hydrate tartrate, dilithium 1+ ion hydrate tartrate, lithium 2,3-dihydroxybutanedioate-water 2/1/1 | |
GNDOSKKVQUGWFB-UHFFFAOYSA-L | |
dilithium;2,3-dihydroxybutanedioate;hydrate | |
18463206 | |
99% |
6108-32-3 | |
C4H6Li2O7 | |
250 g | |
Soluble in water | |
[Li+].[Li+].C(C(C(=O)[O-])O)(C(=O)[O-])O.O | |
179.965 | |
179.97 (161.97 Anhydrous) | |
Powder or Aggregates |
Chemical Identifiers
6108-32-3 | |
179.965 | |
GNDOSKKVQUGWFB-UHFFFAOYSA-L | |
18463206 | |
[Li+].[Li+].C(C(C(=O)[O-])O)(C(=O)[O-])O.O |
C4H6Li2O7 | |
MFCD00054421 | |
lithium tartrate monohydrate, lithium tartrate 1-hydrate, c4h4o6.2li.h2o, dilithium 1+ hydrate tartrate, dilithium 1+ ion hydrate tartrate, lithium 2,3-dihydroxybutanedioate-water 2/1/1 | |
dilithium;2,3-dihydroxybutanedioate;hydrate |
Safety and Handling
EINECSNumber : 212-772-8
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only