Please login to your online account to display your discounted pricing

Alfa Aesar™ Lithium tantalum isopropoxide, 99.9% (metals basis)

Chemical Identifiers

Chemical Name or Material Lithium tantalum isopropoxide
CAS 21864-24-4
Linear Formula LiTa[OCH(CH3)2]6
Formula Weight 542.42
MDL Number MFCD00798538
UN Number UN2925
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4052004

  SDS
alfa aesar™
4052004
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4052009

  SDS
alfa aesar™
4052009
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4052018

  SDS
alfa aesar™
4052018
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 542.42
Chemical Name or Material Lithium tantalum isopropoxide
Physical Form Wet Solid
Assay 99.9%
Assay Percent Notes (metals basis)
Linear Formula LiTa[OCH(CH3)2]6
MDL Number MFCD00798538
CAS 21864-24-4
Quantity 2g
UN Number UN2925
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H228
Causes severe skin burns and eye damage.
Flammable solid.
Boiling Point 160° to 180°C (0.1mmHg)
Health Hazard 3 GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P310
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Color Off-white
DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, CORROSIVE, ORGANIC, N.O.S.
Sensitivity Moisture Sensitive
Safety and Handling

Danger

GHS P Statement
P210-P280-P303+P361+P353-P305+P351+P338-P310
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H228
Causes severe skin burns and eye damage.
Flammable solid.