Please login to your online account to display your discounted pricing

Alfa Aesar™ Lithium tantalum ethoxide, 99.9% (metals basis), 5% w/v in ethanol

Precursor for Lithium tantalum oxide films

Chemical Identifiers

Chemical Name or Material Lithium tantalum ethoxide
CAS 127503-04-2
Linear Formula LiTa(OCH2CH3)6
Formula Weight 458.04
MDL Number MFCD00210630
UN Number UN1170
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA42344AC

 
alfa aesar™
42344AC
25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA42344AE

 
alfa aesar™
42344AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 458.04
Chemical Name or Material Lithium tantalum ethoxide
Health Hazard 1 Danger
Color Pale yellow
Concentration 5% w/v in ethanol
Physical Form Liquid
Assay 99.9%
Assay Percent Notes (metals basis)
Linear Formula LiTa(OCH2CH3)6
MDL Number MFCD00210630
CAS 127503-04-2
Quantity 25mL
UN Number UN1170
Health Hazard 2 GHS H Statement
H225
Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Odor Alcohol-like
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHANOL SOLUTIONS
Sensitivity Moisture Sensitive
Safety and Handling

Danger

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225
Highly flammable liquid and vapor.