Please login to your online account to display your discounted pricing

Lithium sulfate monohydrate, 99%, Acros Organics


Quantity 100g

Manufacturer: thermo fisher scientific  AC447911000

 View more versions of this product

Catalog No. AC447911000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Lithium sulfate monohydrate
CAS 10102-25-7
Purity Grade For biochemistry
Assay Percent Range 99%
Molecular Formula Li2O4S·H2O
Formula Weight 127.96
Packaging Plastic bottle
Quantity 100g

Specifications

Chemical Name or Material Lithium sulfate monohydrate
CAS 10102-25-7
Purity Grade For biochemistry
Assay Percent Range 99%
Molecular Formula Li2O4S·H2O
Formula Weight 127.96