Please login to your online account to display your discounted pricing

Lithium sulfate monohydrate, 99%, Acros Organics

$418.65 - $418.65

Chemical Identifiers

Chemical Name or Material Lithium sulfate monohydrate
CAS 10102-25-7
Purity Grade For biochemistry
Assay Percent Range 99%
Molecular Formula Li2O4S·H2O
Formula Weight 127.96
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC447915000

  SDS
thermo fisher scientific
AC447915000
500g Each for $418.65

AC447911000

  SDS
thermo fisher scientific
AC447911000
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 127.96
Quantity 500g
Purity Grade For biochemistry
Chemical Name or Material Lithium sulfate monohydrate
Packaging Plastic bottle
Assay Percent Range 99%
Molecular Formula Li2O4S·H2O
CAS 10102-25-7