Learn More
Lithium sulfate monohydrate, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1041022
Description
Lithium sulfate is used to treat bipolar disorder. It is also used in ultrasound-type non-destructive testing because they are very efficient sound generators. It is also used in building blocks and chemical synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsLithium sulfate is used to treat bipolar disorder. It is also used in ultrasound-type non-destructive testing because they are very efficient sound generators. It is also used in building blocks and chemical synthesis.
Solubility
Soluble in water. 340 g/L (20 C)
Notes
Store under dry inert gas. Protect from humidity and water. Hygroscopic. Incompatible with strong oxidizing agents.
Specifications
Lithium sulfate monohydrate | |
10102-25-7 | |
H2Li2O5S | |
14,5541 | |
Soluble in water. 340g/L (20 C) | |
[Li+].[Li+].O.[O-]S([O-])(=O)=O | |
127.95 | |
127.96 (109.94 Anhydrous) | |
Odorless | |
2.06 |
130°C (in -H2O) | |
100 g | |
MFCD00149766 | |
lithium sulfate monohydrate, dilithium 1+ ion hydrate sulfate, dilithium sulfate hydrate, sufuric acid, dilithium slat, monohydrate, lithiumsulfatemonohydrate, dilithotab hydrate sulfate, lithium sulphate monohydrate, lithium sulfate, acs, ksc181s7n | |
RKGLUDFWIKNKMX-UHFFFAOYSA-L | |
dilithium;sulfate;hydrate | |
165815 | |
99% | |
Hygroscopic |
Chemical Identifiers
10102-25-7 | |
127.95 | |
RKGLUDFWIKNKMX-UHFFFAOYSA-L | |
165815 | |
[Li+].[Li+].O.[O-]S([O-])(=O)=O |
H2Li2O5S | |
MFCD00149766 | |
lithium sulfate monohydrate, dilithium 1+ ion hydrate sulfate, dilithium sulfate hydrate, sufuric acid, dilithium slat, monohydrate, lithiumsulfatemonohydrate, dilithotab hydrate sulfate, lithium sulphate monohydrate, lithium sulfate, acs, ksc181s7n | |
dilithium;sulfate;hydrate |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280i-P301+P312-P304+P340-P305+P351+P338-P312-P330-P501c
H302-H319-H335
EINECSNumber : 233-820-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only