missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lithium sulfate hydrate, Puratronic™, 99.996% (metals basis)

Manufacturer:  Alfa Aesar™ 1298306

 View more versions of this product

Catalog No. AA1298306



Specifications

Specifications

Lithium sulfate hydrate
(metals basis)
Li2SO4·xH2O
14,5541
INHCSSUBVCNVSK-UHFFFAOYSA-L
dilithium;sulfate
66320
109.94 (Anhydrous)
99.996%
2.06g/mL
130°C (in -H2O)
10377-48-7
Li2O4S
MFCD00011086
lithium sulfate, lithium sulphate, dilithium sulfate, sulfuric acid, dilithium salt, lithiophor, li2so4, dilthium sulfate, sulfuric acid, lithium salt 1:2, unii-919xa137jk, lithium sulfate, anhydrous
[Li+].[Li+].[O-]S(=O)(=O)[O-]
109.936
CHEBI:53474
Crystalline
Puratronic™
White
5g

Chemical Identifiers

10377-48-7
109.936
INHCSSUBVCNVSK-UHFFFAOYSA-L
66320
dilithium;sulfate
Li2O4S
MFCD00011086
lithium sulfate, lithium sulphate, dilithium sulfate, sulfuric acid, dilithium salt, lithiophor, li2so4, dilthium sulfate, sulfuric acid, lithium salt 1:2, unii-919xa137jk, lithium sulfate, anhydrous
CHEBI:53474
[Li+].[Li+].[O-]S(=O)(=O)[O-]
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : OJ6419000

TSCA : TSCA

Documents