Learn More
Alfa Aesar™ Lithium sulfate hydrate, Puratronic™, 99.996% (metals basis)
Manufacturer: Alfa Aesar™ 1298314
Specifications
Lithium sulfate hydrate | |
(metals basis) | |
Li2SO4·xH2O | |
14,5541 | |
INHCSSUBVCNVSK-UHFFFAOYSA-L | |
dilithium;sulfate | |
66320 | |
109.94 (Anhydrous) | |
99.996% | |
2.06g/mL | |
130°C (in -H2O) |
10377-48-7 | |
Li2O4S | |
MFCD00011086 | |
lithium sulfate, lithium sulphate, dilithium sulfate, sulfuric acid, dilithium salt, lithiophor, li2so4, dilthium sulfate, sulfuric acid, lithium salt 1:2, unii-919xa137jk, lithium sulfate, anhydrous | |
[Li+].[Li+].[O-]S(=O)(=O)[O-] | |
109.936 | |
CHEBI:53474 | |
Crystalline | |
Puratronic™ | |
White | |
25g |
Chemical Identifiers
10377-48-7 | |
109.936 | |
INHCSSUBVCNVSK-UHFFFAOYSA-L | |
66320 | |
dilithium;sulfate |
Li2O4S | |
MFCD00011086 | |
lithium sulfate, lithium sulphate, dilithium sulfate, sulfuric acid, dilithium salt, lithiophor, li2so4, dilthium sulfate, sulfuric acid, lithium salt 1:2, unii-919xa137jk, lithium sulfate, anhydrous | |
CHEBI:53474 | |
[Li+].[Li+].[O-]S(=O)(=O)[O-] |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
RTECSNumber : OJ6419000
TSCA : TSCA