missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lithium sulfate, anhydrous, 99.99% (metals basis)

Manufacturer:  Alfa Aesar™ 4435222

 View more versions of this product

Catalog No. AA4435222



Specifications

Specifications

Lithium sulfate
10377-48-7
Li2O4S
14,5541
Soluble in water(345g/L). Insoluble in absolute ethanol,acetone and pyridine.
[Li+].[Li+].[O-]S(=O)(=O)[O-]
109.936
CHEBI:53474
Coarse Powder
2.22 g/mL
Hygroscopic
845°C
Anhydrous
(metals basis)
MFCD00011086
dilithium sulfate
INHCSSUBVCNVSK-UHFFFAOYSA-L
dilithium;sulfate
66320
109.94
99.99%
Odorless
White
100g

Chemical Identifiers

10377-48-7
109.936
INHCSSUBVCNVSK-UHFFFAOYSA-L
66320
dilithium;sulfate
Li2O4S
MFCD00011086
dilithium sulfate
CHEBI:53474
[Li+].[Li+].[O-]S(=O)(=O)[O-]
Safety and Handling

Safety and Handling

GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.

P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c

H302-H319

EINECSNumber : 233-820-4

RTECSNumber : OJ6419000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

Documents