missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium sulfate, anhydrous, 99.7% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1340430

 View more versions of this product

Catalog No. AA1340430



Specifications

Specifications

Lithium sulfate, anhydrous
99.7% (Metal basis)
MFCD00011086
lithium sulfate, lithium sulphate, dilithium sulfate, sulfuric acid, dilithium salt, lithiophor, li2so4, dilthium sulfate, sulfuric acid, lithium salt 1:2, unii-919xa137jk, lithium sulfate, anhydrous
[Li+].[Li+].[O-]S(=O)(=O)[O-]
109.936
CHEBI:53474
Powder
Odorless
250g
10377-48-7
Li2O4S
White powder
INHCSSUBVCNVSK-UHFFFAOYSA-L
dilithium;sulfate
66320
109.94
2.22g/cm3 at 20°C
White

Chemical Identifiers

10377-48-7
109.936
INHCSSUBVCNVSK-UHFFFAOYSA-L
66320
dilithium;sulfate
Li2O4S
MFCD00011086
lithium sulfate, lithium sulphate, dilithium sulfate, sulfuric acid, dilithium salt, lithiophor, li2so4, dilthium sulfate, sulfuric acid, lithium salt 1:2, unii-919xa137jk, lithium sulfate, anhydrous
CHEBI:53474
[Li+].[Li+].[O-]S(=O)(=O)[O-]
Safety and Handling

Safety and Handling

H302: Harmful if swallowed.

EINECSNumber : 233-820-4

Documents