Please login to your online account to display your discounted pricing

Lithium, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical


For atomic absorption. (1mL = 1mg Li) Solute: Lithium Carbonate Solvent: Dilute Hydrochloric Acid

Quantity 500mL
Packaging Poly Bottle

Manufacturer: fisher scientific company  FLSL45500

 View more versions of this product

Catalog No. SL45-500

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Boiling Point 100°C
Purity Grade Certified Grade
Melting Point 0°C
Quantity 500mL
DOT Information DOT Class 8, : Corrosive
Physical Form Liquid
pH 2.0
Color Colorless
Vapor Pressure 14mmHg
Packaging Poly Bottle
CAS 7732-18-5
ChemAlert Storage Symbol White
Chemical Name or Material Lithium, Reference Standard Solution
CAS 554-13-2
CAS 7647-01-0
MDL Number MFCD00134051
Concentration or Composition (by Analyte or Components) 1000ppm ±1%
Traceability to NIST LOT NUMBER USED lithium ref. Std.
Concentration 990 to 1010ppm
Name Note 1000ppm ±1%

Specifications

Chemical Name or Material Lithium, Reference Standard Solution
Name Note 1000ppm ±1%
CAS 554-13-2
CAS 7647-01-0
CAS 7732-18-5
Purity Grade Certified Grade
MDL Number MFCD00134051
Accessories
 

SDS & Safety Documents