Please login to your online account to display your discounted pricing

Lithium, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical

For atomic absorption. (1mL = 1mg Li) Solute: Lithium Carbonate Solvent: Dilute Hydrochloric Acid

$78.88 - $80.88

Chemical Identifiers

Chemical Name or Material Lithium, Reference Standard Solution
Name Note 1000ppm ±1%
CAS 554-13-2
CAS 7647-01-0
CAS 7732-18-5
Purity Grade Certified Grade
MDL Number MFCD00134051
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SL45-100

  SDS
fisher scientific company
SL45100
Poly Bottle 100mL Each for $80.88
Case of 6 Each for $485.28

SL45-500

  SDS
fisher scientific company
FLSL45500
Poly Bottle 500mL Each for $78.88
Case of 6 Each for $473.28
Description & Specifications

Specifications

Boiling Point 100°C
Melting Point 0°C
Purity Grade Certified Grade
Quantity 100mL
DOT Information DOT Class 8, : Corrosive
Physical Form Liquid
Vapor Pressure 14mmHg
pH 2.0
Color Colorless
CAS 7647-01-0
CAS 7732-18-5
Name Note 1000ppm ±1%
Chemical Name or Material Lithium, Reference Standard Solution
MDL Number MFCD00134051
Concentration or Composition (by Analyte or Components) 1000ppm ±1%
Packaging Poly Bottle
Traceability to NIST LOT NUMBER USED lithium ref. Std.
Concentration 990 to 1010ppm
ChemAlert Storage Symbol White
CAS 554-13-2
Safety and Handling