Please login to your online account to display your discounted pricing

Alfa Aesar™ Lithium pyruvate monohydrate

$58.31 - $230.54

Chemical Identifiers

Chemical Name or Material Lithium pyruvate monohydrate
CAS 2922-61-4
Molecular Formula CH3C(O)CO2Li·H2O
Formula Weight 112.01 (94.00 Anhydrous)
MDL Number MFCD00150352
Synonym Pyruvic acid lithium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1630809

 
alfa aesar™
A1630809
10g Each for $58.31

AAA1630818

 
alfa aesar™
A1630818
50g Each for $230.54
Description & Specifications

Specifications

Formula Weight 112.01 (94.00 Anhydrous)
Chemical Name or Material Lithium pyruvate monohydrate
CAS 2922-61-4
Molecular Formula CH3C(O)CO2Li·H2O
MDL Number MFCD00150352
Synonym Pyruvic acid lithium salt
TSCA TSCA
Quantity 10g