missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium plasma standard solution, Alfa Aesar™, Specpure™

Manufacturer:  Alfa Aesar 13821AP

 View more versions of this product

Catalog No. AA13821AP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Lithium
Clear liquid
MFCD00134051
XGZVUEUWXADBQD-UHFFFAOYSA-L
dilithium;carbonate
11125
Liquid
Colorless
554-13-2,7697-37-2,7732-18-5
CLi2O3
Nitric acid: 5.0%; Lithium carbonate: 0.53%; Water: 94.47%
[Li+].[Li+].C(=O)([O-])[O-]
73.888
CHEBI:6504
23 hPa (17mm Hg) at 20°C
500mL

Chemical Identifiers

554-13-2
73.888
XGZVUEUWXADBQD-UHFFFAOYSA-L
CHEBI:6504
[Li+].[Li+].C(=O)([O-])[O-]
CLi2O3
MFCD00134051
11125
dilithium;carbonate
Safety and Handling

Safety and Handling

H314: Causes severe skin burns and eye damage.

DOTInformation : Hazard Class: 8, Packing Group: III

Documents