Please login to your online account to display your discounted pricing

Alfa Aesar™ Lithium phthalocyanine

$99.43 - $99.43

Chemical Identifiers

Chemical Name or Material Lithium phthalocyanine
CAS 25510-41-2
Formula Weight 526.41
MDL Number MFCD00059094
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3933406

  SDS
alfa aesar™
3933406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3933403

  SDS
alfa aesar™
3933403
1g Each for $99.43
Description & Specifications

Specifications

Formula Weight 526.41
Chemical Name or Material Lithium phthalocyanine
TSCA TSCA
Physical Form Powder
MDL Number MFCD00059094
CAS 25510-41-2
Quantity 5g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.