missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium phosphate, Puratronic™, 99.99% (metals basis), Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 1074314

 View more versions of this product

Catalog No. AA1074314



Specifications

Specifications

Lithium phosphate
99.99% (Metals basis)
Li3O4P
li3po4
[Li+].[Li+].[Li+].[O-]P(=O)([O-])[O-]
115.79
115.79
99.99%
Odorless
25g
10377-52-3
(metals basis)
MFCD00016187
TWQULNDIKKJZPH-UHFFFAOYSA-K
trilithium;phosphate
165867
Powder
Puratronic™
1205°C

Chemical Identifiers

10377-52-3
115.79
TWQULNDIKKJZPH-UHFFFAOYSA-K
165867
[Li+].[Li+].[Li+].[O-]P(=O)([O-])[O-]
Li3O4P
MFCD00016187
li3po4
trilithium;phosphate
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA

Documents