missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lithium peroxide, 95%

Manufacturer:  Alfa Aesar™ 1342122

 View more versions of this product

Catalog No. AA1342122



Specifications

Specifications

Lithium peroxide
Li2O2
UN1472
HPGPEWYJWRWDTP-UHFFFAOYSA-N
dilithium;peroxide
25489
Powder
Odorless
White
12031-80-0
MFCD00016184
dilithium peroxide, lithium peroxide li2 o2, lithium oxide, unii-9anx556r5f, lithium peroxide un1472 oxidizer
[Li+].[Li+].[O-][O-]
45.878
45.88
95%
Air & Moisture Sensitive
100g

Chemical Identifiers

12031-80-0
45.878
HPGPEWYJWRWDTP-UHFFFAOYSA-N
25489
[Li+].[Li+].[O-][O-]
Li2O2
MFCD00016184
dilithium peroxide, lithium peroxide li2 o2, lithium oxide, unii-9anx556r5f, lithium peroxide un1472 oxidizer
dilithium;peroxide
Safety and Handling

Safety and Handling

GHS H Statement
H272-H314-H318
May intensify fire
oxidizer.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P221-P210-P303+P361+P353-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: LITHIUM PEROXIDE

TSCA : TSCA

Documents