Learn More
Lithium perchlorate trihydrate, ACS, 63.0-68.0% LiClO{4}, Thermo Scientific Chemicals
$119.32 - $325.45
Chemical Identifiers
CAS | 13453-78-6 |
---|---|
Molecular Formula | ClH6LiO7 |
Molecular Weight (g/mol) | 160.431 |
MDL Number | MFCD00149765 |
InChI Key | JAWYRNYHJJDXHX-UHFFFAOYSA-M |
Synonym | lithium perchlorate trihydrate, perchloric acid, lithium salt, trihydrate, unii-jyk745743v, acmc-1c1zx, lithium perchlorate hydrate, lithiumperchlorate trihydrate, ksc491a4p, lithium pechlorate trihydrate, lithiumperchloratetrihydrate, lithotab trihydrate perchlorate ion |
PubChem CID | 23708903 |
IUPAC Name | lithium;perchlorate;trihydrate |
SMILES | [Li+].O.O.O.[O-]Cl(=O)(=O)=O |
Description
Lithium perchlorate trihydrate is used as an oxygen source for chemical oxygen generators as it liberates oxygen at high temperatures. Excellent solubility of lithium perchlorate permits its use in Diels-Alder reactions. It is also employed in lithium-ion batteries as an electrolyte and acts as a chaotropic agent to denature proteins. It is used in the Baylis-Hillman reaction as a catalyst in the coupling of alpha, beta-unsaturated carbonyl compounds with aldehydes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13453-78-6 | |
160.431 | |
JAWYRNYHJJDXHX-UHFFFAOYSA-M | |
23708903 | |
[Li+].O.O.O.[O-]Cl(=O)(=O)=O |
ClH6LiO7 | |
MFCD00149765 | |
lithium perchlorate trihydrate, perchloric acid, lithium salt, trihydrate, unii-jyk745743v, acmc-1c1zx, lithium perchlorate hydrate, lithiumperchlorate trihydrate, ksc491a4p, lithium pechlorate trihydrate, lithiumperchloratetrihydrate, lithotab trihydrate perchlorate ion | |
lithium;perchlorate;trihydrate |
Specifications
13453-78-6 | |
Crystalline | |
ClH6LiO7 | |
MFCD00149765 | |
14,5539 | |
Highly soluble in water. | |
[Li+].O.O.O.[O-]Cl(=O)(=O)=O | |
160.431 | |
160.45 (106.40 Anhydrous) | |
63.0 to 68.0% LiClO4 | |
1.84 g/mL |
White | |
100 g | |
LiClO4·3H2O | |
UN1481 | |
lithium perchlorate trihydrate, perchloric acid, lithium salt, trihydrate, unii-jyk745743v, acmc-1c1zx, lithium perchlorate hydrate, lithiumperchlorate trihydrate, ksc491a4p, lithium pechlorate trihydrate, lithiumperchloratetrihydrate, lithotab trihydrate perchlorate ion | |
JAWYRNYHJJDXHX-UHFFFAOYSA-M | |
lithium;perchlorate;trihydrate | |
23708903 | |
ACS | |
Odorless | |
Lithium perchlorate trihydrate |
Safety and Handling
GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P380+P375-P372-P380-P501c
H272-H315-H319-H335
DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.
EINECSNumber : 232-237-2
RTECSNumber : SC8750000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only