missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lithium perchlorate trihydrate, ACS, 63.0-68.0% LiClO4

Manufacturer:  Alfa Aesar™ 4592322

 View more versions of this product

Catalog No. AA4592322



Specifications

Specifications

Lithium perchlorate trihydrate
ClH6LiO7
MFCD00149765
14,5539
JAWYRNYHJJDXHX-UHFFFAOYSA-M
lithium;perchlorate;trihydrate
23708903
Crystalline
63.0 to 68.0% LiClO4
Odorless
100g
13453-78-6
LiClO4·3H2O
UN1481
lithium perchlorate trihydrate, perchloric acid, lithium salt, trihydrate, unii-jyk745743v, acmc-1c1zx, lithium perchlorate hydrate, lithiumperchlorate trihydrate, ksc491a4p, lithium pechlorate trihydrate, lithiumperchloratetrihydrate, lithotab trihydrate perchlorate ion
[Li+].O.O.O.[O-]Cl(=O)(=O)=O
160.431
160.45 (106.40 Anhydrous)
ACS
1.84g/mL
White

Chemical Identifiers

13453-78-6
160.431
JAWYRNYHJJDXHX-UHFFFAOYSA-M
23708903
[Li+].O.O.O.[O-]Cl(=O)(=O)=O
ClH6LiO7
MFCD00149765
lithium perchlorate trihydrate, perchloric acid, lithium salt, trihydrate, unii-jyk745743v, acmc-1c1zx, lithium perchlorate hydrate, lithiumperchlorate trihydrate, ksc491a4p, lithium pechlorate trihydrate, lithiumperchloratetrihydrate, lithotab trihydrate perchlorate ion
lithium;perchlorate;trihydrate
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.

RTECSNumber : SC8750000

TSCA : TSCA

SDS Documents