Learn More
Lithium perchlorate trihydrate, 99%, extra pure, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 194720250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Oxidation reagentSpecifications
Lithium perchlorate trihydrate | |
13453-78-6 | |
98.5% min. (Argentometry) | |
LiClO4·3H2O | |
15, 5595 | |
Solubility in water: 597g/L water (25°C). Other solubilities: soluble in alcohol | |
[Li+].O.O.O.[O-]Cl(=O)(=O)=O | |
160.44 | |
160.44 | |
Extra Pure |
p.a. | |
25g | |
ClLiO4·3H2O | |
MFCD00149765 | |
lithium perchlorate trihydrate, perchloric acid, lithium salt, trihydrate, unii-jyk745743v, acmc-1c1zx, lithium perchlorate hydrate, lithiumperchlorate trihydrate, ksc491a4p, lithium pechlorate trihydrate, lithiumperchloratetrihydrate, lithotab trihydrate perchlorate ion | |
JAWYRNYHJJDXHX-UHFFFAOYSA-M | |
lithium;perchlorate;trihydrate | |
23708903 | |
99% | |
Plastic bottle |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May intensify fire; oxidizer.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
RTECSNumber : SC8750000
RUO â Research Use Only