missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lithium perchlorate, anhydrous, 99.99% (metals basis)

Manufacturer:  Alfa Aesar™ 4422618

 View more versions of this product

Catalog No. AA4422618

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

  • Ampouled under argon
Specifications

Specifications

Lithium perchlorate
7791-03-9
ClH6LiO7
UN1481
lithium perchlorate, perchloric acid, lithium salt, lithiumperchlorate, unii-q86se98c9c, lithium 1+ ion perchlorate, lithium perchlorate, anhydrous, lithium cloricum, lithium per chlorate, liclo4, perchloric acid, lithium salt 1:1
[Li+].[O-]Cl(=O)(=O)=O
160.43
106.4
99.99%
Ampule
400°C (decomposition)
236°C
Anhydrous
(metals basis)
MFCD00011079
14,5539
JAWYRNYHJJDXHX-UHFFFAOYSA-M
lithium;perchlorate
23665649
Lumps
2.43g/mL
Hygroscopic
White
50g

Chemical Identifiers

7791-03-9
160.43
JAWYRNYHJJDXHX-UHFFFAOYSA-M
23665649
[Li+].[O-]Cl(=O)(=O)=O
ClH6LiO7
MFCD00011079
lithium perchlorate, perchloric acid, lithium salt, lithiumperchlorate, unii-q86se98c9c, lithium 1+ ion perchlorate, lithium perchlorate, anhydrous, lithium cloricum, lithium per chlorate, liclo4, perchloric acid, lithium salt 1:1
lithium;perchlorate
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.

TSCA : TSCA

storageNote1 :

  • Ampouled under argon

SDS Documents