missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium oxalate, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 01342618

 View more versions of this product

Catalog No. AA1342618



Description

Description

Lithium oxalate is used as lithium battery electrolyte. It is also used as an analytical reagent and reducing reagent. Further, it is used as an intermediate in the manufacture of other chemicals such as pharmaceuticals, agrochemicals, dyestuffs and in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Lithium oxalate
(decomposition)
C2Li2O4
50 g
14,5537
Soluble in water.
[Li+].[Li+].[O-]C(=O)C([O-])=O
101.90
101.96
Powder
553-91-3
2.12 g/mL
MFCD00040596
UN3288
lithium oxalate, dilithium oxalate, ethanedioic acid, dilithium salt, unii-k737ot0e73, oxalic acid dilithium salt, ethanedioic acid, lithium salt 1:2, oxalic acid dilithium, acmc-1ctwf, c2o4.2li, ksc492a1t
YNQRWVCLAIUHHI-UHFFFAOYSA-L
dilithium;oxalate
68383
≥99%

Chemical Identifiers

553-91-3
101.90
YNQRWVCLAIUHHI-UHFFFAOYSA-L
68383
C2Li2O4
MFCD00040596
lithium oxalate, dilithium oxalate, ethanedioic acid, dilithium salt, unii-k737ot0e73, oxalic acid dilithium salt, ethanedioic acid, lithium salt 1:2, oxalic acid dilithium, acmc-1ctwf, c2o4.2li, ksc492a1t
[Li+].[Li+].[O-]C(=O)C([O-])=O
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.

P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c

H302+H312

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

EINECSNumber : 209-054-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only