missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lithium oxalate, 99+%

Manufacturer:  Alfa Aesar™ 1342630

 View more versions of this product

Catalog No. AA1342630



Specifications

Specifications

Lithium oxalate
250g
C2Li2O4
UN3288
lithium oxalate, dilithium oxalate, ethanedioic acid, dilithium salt, unii-k737ot0e73, oxalic acid dilithium salt, ethanedioic acid, lithium salt 1:2, oxalic acid dilithium, acmc-1ctwf, c2o4.2li, ksc492a1t
YNQRWVCLAIUHHI-UHFFFAOYSA-L
dilithium;oxalate
68383
Powder
553-91-3
2.12g/mL
MFCD00040596
14,5537
Soluble in water
[Li+].[Li+].C(=O)(C(=O)[O-])[O-]
101.898
101.96
≥99%

Chemical Identifiers

553-91-3
101.898
YNQRWVCLAIUHHI-UHFFFAOYSA-L
68383
[Li+].[Li+].C(=O)(C(=O)[O-])[O-]
C2Li2O4
MFCD00040596
lithium oxalate, dilithium oxalate, ethanedioic acid, dilithium salt, unii-k737ot0e73, oxalic acid dilithium salt, ethanedioic acid, lithium salt 1:2, oxalic acid dilithium, acmc-1ctwf, c2o4.2li, ksc492a1t
dilithium;oxalate
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P280-P301+P312-P312-P363-P322-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

TSCA : TSCA

Documents