Please login to your online account to display your discounted pricing

Lithium Nitrate, Reagent Grade, LabChem™

$300.29 - $300.29

Chemical Identifiers

Chemical Name or Material Lithium Nitrate
CAS 7790-69-4
CAS Max % 100
Purity Grade Reagent Grade
Linear Formula LiNO3
Formula Weight 68.96
UN Number UN2722
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC162501

 
lab chem
LC162501
500g Each for $300.29
Description & Specifications

Specifications

Formula Weight 68.96
Chemical Name or Material Lithium Nitrate
EINECS Number 232-218-9
Identification Passes Test
Packaging Poly Bottle
Purity Grade Reagent Grade
Physical Form Granules
Solubility Soluble in water
Specific Gravity 2380kg/m3
Storage Information Room Temperature
Linear Formula LiNO3
CAS 7790-69-4
UN Number UN2722
Decomposition Information Lithium oxide; Nitrogen oxides
Color Colorless
Melting Point 264°C
Quantity 500g
Health Hazard 1 Warning
CAS Max % 100
Health Hazard 2 GHS H Statement
May intensify fire; oxidizer.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep/Store away from clothing, combustible materials.
Take any precaution to avoid mixing with combustibles.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
If on skin: Take off contaminated clothing.
Wash skin with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Wash contaminated clothing before reuse.
In case of fire: Use carbon dioxide (CO2), powder, alcohol-resistant foam to extinguish.
Dispose of contents/container to comply with local, state and federal regulations.
Density 2,380kg/m3
Safety and Handling

Warning

GHS P Statement
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep/Store away from clothing, combustible materials.
Take any precaution to avoid mixing with combustibles.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
If on skin: Take off contaminated clothing.
Wash skin with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Wash contaminated clothing before reuse.
In case of fire: Use carbon dioxide (CO2), powder, alcohol-resistant foam to extinguish.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
May intensify fire; oxidizer.
Causes skin irritation.
Causes serious eye irritation.