missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lithium nitrate hydrate, 99.999% (metals basis), Alfa Aesar™, Puratronic™

Manufacturer:  Thermo Fisher Scientific 1074218

 View more versions of this product

Catalog No. AA1074218



Specifications

Specifications

Lithium nitrate hydrate
99.99% (metals basis)
MFCD00011094
IIPYXGDZVMZOAP-UHFFFAOYSA-N
lithium;nitrate
10129889
Crystalline Aggregates
7790-69-4
LiNO3
ksc492i7j
[Li+].[N+](=O)([O-])[O-]
68.944
CHEBI:63315
50g

Chemical Identifiers

7790-69-4
68.944
IIPYXGDZVMZOAP-UHFFFAOYSA-N
10129889
lithium;nitrate
LiNO3
MFCD00011094
ksc492i7j
CHEBI:63315
[Li+].[N+](=O)([O-])[O-]
Safety and Handling

Safety and Handling

EINECSNumber : 232-218-9

Documents