Please login to your online account to display your discounted pricing

Lithium Nitrate, Crystal, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$445.30 - $1,595.55

Chemical Identifiers

Chemical Name or Material Lithium Nitrate
CAS 7790-69-4
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula LiNO3
Formula Weight 68.94
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003219

 
avantor performance
238405
Poly Bottle 2.5kg Each for $1,595.55

02003220

 
avantor performance
238401
Poly Bottle 500g Each for $445.30
Description & Specifications

Specifications

Physical Form Crystals
Formula Weight 68.94
Chemical Name or Material Lithium Nitrate
CAS 7790-69-4
Molecular Formula LiNO3
Quantity 2.5kg
Packaging Poly Bottle
Specific Gravity 1L = 2.38kg
Purity Grade BAKER ANALYZED™ Reagent